Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081848.1 | NP_001075317.1 | 1 - 2 | 46 | 87 | |
GenBank mRNA | AF162703.1 | - | 1 - 2 | 46 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |