Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081860.2 | NP_001075329.1 | 2 - 3 | 173 | 105 | |
NM_001309387.1 | NP_001296316.1 | 3 - 4 | 328 | 105 | ||
GenBank mRNA | AF503365.1 | - | 2 - 3 | 189 | 105 | |
BI961242.1 | - | 3 - 4 | 335 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |