Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081868.1 | NP_001075337.1 | 4 - 5 | 481 | 88 | |
GenBank mRNA | AJ133469.1 | - | 4 - 5 | 481 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |