Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081870.2 | NP_001075339.1 | 3 - 4 | 313 | 66 | |
GenBank mRNA | AJ243283.1 | - | 3 - 4 | 313 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |