Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081873.1 | NP_001075342.1 | 5 - 6 | 664 | 90 | |
XM_005603492.3 | XP_005603549.1 | 658 | 90 | |||
GenBank mRNA | AJ006631.1 | - | 3 - 4 | 339 | 90 | |
AJ315379.1 | - | 5 - 6 | 664 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |