Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081874.1 | NP_001075343.1 | 1 - 2 | 60 | 66 | |
XM_014734198.2 | XP_014589684.2 | 112 | 66 | |||
GenBank mRNA | AJ459964.1 | - | 1 - 2 | 60 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |