Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081894.2 | NP_001075363.1 | 4 - 5 | 743 | 93 | |
GenBank mRNA | AY293741.1 | - | 4 - 5 | 743 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |