Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081909.1 | NP_001075378.1 | 1 - 2 | 444 | 62 | |
XM_023638729.1 | XP_023494497.1 | 632 | 62 | |||
GenBank mRNA | D50326.1 | - | 1 - 2 | 444 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |