Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081943.1 | NP_001075412.1 | 6 - 7 | 790 | 69 | |
GenBank mRNA | L38511.1 | - | 6 - 7 | 790 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |