Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082495.2 | NP_001075964.1 | 4 - 5 | 679 | 65 | |
GenBank mRNA | U62529.1 | - | 4 - 5 | 681 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |