Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082500.2 | NP_001075969.2 | 3 - 4 | 156 | 115 | |
GenBank mRNA | CD466534.1 | - | 3 - 4 | 156 | 115 | |
D42146.1 | - | 2 - 3 | 108 | 115 | ||
U92480.1 | - | 3 - 4 | 135 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |