Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082511.2 | NP_001075980.2 | 6 - 7 | 612 | 74 | |
GenBank mRNA | Y11130.1 | - | 510 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Inventoried Probe spans exons |