Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082523.1 | NP_001075992.1 | 1 - 2 | 335 | 79 | |
GenBank mRNA | D30688.1 | - | 1 - 2 | 335 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |