Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083951.2 | NP_001077420.2 | 1 - 2 | 162 | 69 | |
GenBank mRNA | AF062377.1 | - | 1 - 2 | 70 | 69 | |
AY184956.1 | - | 1 - 2 | 165 | 69 | ||
CD536564.1 | - | 1 - 2 | 162 | 69 | ||
JL626338.1 | - | 1 - 2 | 93 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |