Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109680.1 | NP_001103150.1 | 4 - 5 | 627 | 86 | |
XM_023626553.1 | XP_023482321.1 | 1430 | 86 | |||
XM_023626556.1 | XP_023482324.1 | 2151 | 86 | |||
XM_023626563.1 | XP_023482331.1 | 1200 | 86 | |||
GenBank mRNA | EU029660.1 | - | 4 - 5 | 627 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons |