Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122656.1 | NP_001116128.1 | 1 - 2 | 130 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |