Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130486.2 | NP_001123958.1 | 3 - 4 | 528 | 132 | |
GenBank mRNA | EF397508.1 | - | 3 - 4 | 362 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |