Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163879.1 | NP_001157351.1 | 1 - 2 | 124 | 96 | |
GenBank mRNA | AB035518.1 | - | 1 - 2 | 41 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |