Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163879.1 | NP_001157351.1 | 3 - 4 | 394 | 71 | |
GenBank mRNA | AB035518.1 | - | 3 - 4 | 311 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |