Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163889.1 | NP_001157361.1 | 7 - 8 | 1473 | 61 | |
XM_005611061.3 | XP_005611118.1 | 1700 | 61 | |||
XM_023642149.1 | XP_023497917.1 | 1933 | 61 | |||
XM_023642150.1 | XP_023497918.1 | 1780 | 61 | |||
GenBank mRNA | AF076780.1 | - | 3 - 4 | 893 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Equus caballus Probe spans exons |