Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732990.2 | XP_014588476.2 | 346 | 81 | ||
XM_023638775.1 | XP_023494543.1 | 288 | 81 | |||
GenBank mRNA | AF359386.1 | - | 1 - 2 | 159 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |