Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF448228.1 | - | 26 - 27 | 2965 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |