Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163858.2 | NP_001157330.1 | 84 - 85 | 12056 | 157 | |
GenBank mRNA | AF448228.1 | - | 84 - 85 | 11973 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |