Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501450.5 | XP_001501500.1 | 465 | 108 | ||
XM_023654119.1 | XP_023509887.1 | 480 | 108 | |||
GenBank mRNA | AJ318332.1 | - | 3 - 4 | 351 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |