Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242446.1 | NP_001229375.1 | 5 - 6 | 746 | 93 | |
GenBank mRNA | AY246708.1 | - | 3 - 4 | 265 | 93 | |
CD464593.1 | - | 3 - 4 | 271 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |