Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639032.1 | XP_023494800.1 | 771 | 67 | ||
GenBank mRNA | AY752932.1 | - | 2 - 3 | 355 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |