Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164015.1 | NP_001157487.1 | 2 - 3 | 207 | 71 | |
XM_014734209.2 | XP_014589695.1 | 303 | 71 | |||
GenBank mRNA | X74344.1 | - | 195 | 71 | ||
X74503.1 | - | 189 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Probe spans exons |