Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143797.1 | NP_001137269.1 | 6 - 7 | 684 | 67 | |
GenBank mRNA | EF694759.1 | - | 6 - 7 | 684 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |