Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144880.1 | NP_001138352.1 | 5 - 6 | 694 | 76 | |
XM_023644137.1 | XP_023499905.1 | 691 | 76 | |||
GenBank mRNA | AB472335.1 | - | 5 - 6 | 694 | 76 | |
AB477533.1 | - | 5 - 6 | 694 | 76 | ||
GU808337.1 | - | 5 - 6 | 694 | 76 | ||
HQ890012.1 | - | 2 - 3 | 223 | 76 | ||
JL638585.1 | - | 3 - 4 | 416 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |