Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082526.1 | NP_001075995.1 | 4 | 215 | 105 | |
GenBank mRNA | EU438767.1 | - | 3 | 154 | 105 | |
U92481.1 | - | 4 | 215 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Amplicon spans exons and probe does not span exons |