Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163272.1 | NP_001156744.1 | 8 - 9 | 972 | 58 | |
XM_023633191.1 | XP_023488959.1 | 7242 | 58 | |||
XM_023633192.1 | XP_023488960.1 | 7241 | 58 | |||
XM_023633193.1 | XP_023488961.1 | 2523 | 58 | |||
XM_023633194.1 | XP_023488962.1 | 956 | 58 | |||
XM_023633195.1 | XP_023488963.1 | 2523 | 58 | |||
GenBank mRNA | EU554426.2 | - | 8 - 9 | 972 | 58 | |
HQ596523.1 | - | 8 - 9 | 883 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |