Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164011.1 | NP_001157483.1 | 1 - 2 | 189 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |