Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164013.2 | NP_001157485.2 | 1 - 2 | 223 | 62 | |
XM_005599769.3 | XP_005599826.2 | 495 | 62 | |||
XM_023618413.1 | XP_023474181.1 | 512 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Equus caballus Probe spans exons |