Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205192.1 | NP_001192121.1 | 1 - 2 | 79 | 64 | |
XM_023644175.1 | XP_023499943.1 | 130 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |