Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205192.1 | NP_001192121.1 | 4 - 5 | 897 | 85 | |
XM_023644173.1 | XP_023499941.1 | 1438 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |