Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205193.1 | NP_001192122.1 | 2 - 3 | 523 | 114 | |
XM_005611916.3 | XP_005611973.1 | 812 | 114 | |||
XM_005611917.3 | XP_005611974.1 | 961 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |