Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242553.1 | NP_001229482.1 | 2 - 3 | 378 | 73 | |
XM_023630977.1 | XP_023486745.1 | 666 | 73 | |||
XM_023630978.1 | XP_023486746.1 | 790 | 73 | |||
XM_023630979.1 | XP_023486747.1 | 659 | 73 | |||
XM_023630980.1 | XP_023486748.1 | 833 | 73 | |||
XM_023630981.1 | XP_023486749.1 | 719 | 73 | |||
XM_023630982.1 | XP_023486750.1 | 721 | 73 | |||
XM_023630984.1 | XP_023486752.1 | 497 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |