Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245905.1 | NP_001232834.1 | 14 - 15 | 1694 | 71 | |
XM_023618423.1 | XP_023474191.1 | 1788 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |