Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166452.1 | NP_001159924.1 | 3 - 4 | 689 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons |