Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168391.1 | NP_001161863.1 | 3 - 4 | 383 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |