Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 15 - 16 | 681 | 101 | |
XM_014738515.2 | XP_014594001.1 | 692 | 101 | |||
GenBank mRNA | GU196267.1 | - | 15 - 16 | 681 | 101 | |
KP658381.1 | - | 16 - 17 | 688 | 101 | ||
KP658382.1 | - | 14 - 15 | 637 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |