Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 2 - 3 | 98 | 95 | |
GenBank mRNA | GU196267.1 | - | 2 - 3 | 98 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |