Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185087.3 | NP_001172016.3 | 8 - 9 | 1611 | 91 | |
NM_001185088.2 | NP_001172017.2 | 8 - 9 | 1611 | 91 | ||
NM_001185089.2 | NP_001172018.2 | 8 - 9 | 1127 | 91 | ||
XM_005607780.3 | XP_005607837.1 | 1700 | 91 | |||
XM_005607783.3 | XP_005607840.2 | 1696 | 91 | |||
XM_023626768.1 | XP_023482536.1 | 1698 | 91 | |||
XM_023626774.1 | XP_023482542.1 | 1698 | 91 | |||
XM_023626782.1 | XP_023482550.1 | 1667 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |