Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193658.2 | NP_001180587.1 | 4 - 5 | 927 | 76 | |
GenBank mRNA | HM641824.1 | - | 4 - 5 | 927 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Equus caballus Probe spans exons |