Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197039.2 | NP_001183968.1 | 11 - 12 | 1326 | 108 | |
GenBank mRNA | BI961798.1 | - | 2 - 3 | 174 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |