Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204234.1 | NP_001191163.1 | 10 - 11 | 879 | 61 | |
XM_014733464.2 | XP_014588950.1 | 1892 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |