Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204845.1 | NP_001191774.1 | 5 - 6 | 434 | 86 | |
XM_005599777.3 | XP_005599834.1 | 399 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |