Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253841.1 | NP_001240770.1 | 2 - 3 | 289 | 63 | |
XM_014727793.2 | XP_014583279.1 | 184 | 63 | |||
XM_023646584.1 | XP_023502352.1 | 531 | 63 | |||
XM_023646586.1 | XP_023502354.1 | 552 | 63 | |||
GenBank mRNA | HQ890053.1 | - | 2 - 3 | 289 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |