Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364902.1 | NP_001351831.1 | 4 - 5 | 464 | 76 | |
GenBank mRNA | AJ271386.1 | - | 3 - 4 | 460 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |