Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364902.1 | NP_001351831.1 | 9 - 10 | 1339 | 112 | |
GenBank mRNA | AJ271386.1 | - | 8 - 9 | 1335 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |